missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-Well Half-Skirt PCR Microplates

Catalog No. 14223338
Change view
Click to view available options
Color:
Orange
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14223338 Red
14223337 Orange
14223341 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14223338 Supplier Axygen™ Supplier No. PCR96M2HSR

Half-skirt amplification plate with single notch

PCR™ products specifically designed for use within PCR thermal transfer and real time PCR applications. A variety of colors are offered to help organize multi-step PCR processes.

  • 96-well PCR half-skirt amplification plate with a single notch
  • Working volume is 300μL
  • Available in a variety of colors
  • Sold 10 plates/unit; 5 units/Cs.

Specifications

Certifications/Compliance RNase, DNase-free
Format Standard
Array 8 x 12
Color Red
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR thermal transfer, RT-PCR applications
Quantity 50/Cs.
Well Color Clear
Show More Show Less