missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 96-Well Clear V-Bottom 600 μL Polypropylene Deep Well Plate, Sterile, Individually Wrapped
GREENER_CHOICE

Catalog No. 14223345
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14223345 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14223345 Supplier Axygen™ Supplier No. PDW500CSI
  • ONLINE EXCLUSIVE PRICE
    $597.07
    $231.50 / Case of 25 Each

    Save $365.57

    61.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Polypropylene 96 deep well rectangular plates; specially designed for high throughput screening, sample collection, compound storage, compound libraries, and combinatorial chemistry applications.

  • Polypropylene 96 deep well rectangular plates; specially designed for high throughput screening, sample collection, compound storage, compound libraries, and combinatorial chemistry applications
  • Volume: 600 μL
  • Color: Clear
  • Well Shape: Round; SBS Footprint
  • Pre-sterilized; Individually Wrapped
  • Certified RNase, DNase, and Pyrogen Free

Specifications

Certifications/Compliance ANSI, SLAS 1-2004
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Autoclavable
Color Clear
Material Polypropylene
Volume (Metric) Well 600 μL
No. of Wells 96
Sterility Sterile
Well Shape Round
For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
Max. RCF 4000 x g
No. per Case 25
No. per Pack 5
Quantity 25/Cs.
Show More Show Less