missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 384-well Skirted PCR Microplates
Description
Axygen™ 384 well PCR microplate, white, is designed to fit Roche Light Cycler 480. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Includes Ultra Clear sealing film and printed alphanumeric grid for ease of well identification.
RNase-/DNase-free Nonpyrogenic Nonsterile Manufactured with 99.9% pure virgin polypropylene Includes Sealing Film
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | RNase, DNase-free |
| Format | Standard |
| Lid | Without Lid |
| Description | Sealing film |
| Array | 16 x 24 |
| Color | White |
| For Use With (Equipment) | For Roche light cycler 480 |
| Skirt Style | Full |
| Material | Polypropylene |
| Show More |