missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 384-well Skirted PCR Microplates
GREENER_CHOICE

Catalog No. 14222306
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14222306 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222306 Supplier Axygen™ Supplier No. PCR384C

RNase-/DNase-free

Axygen™ 384 well skirted PCR microplate, clear, is designed to fit standard 384 well blocks. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Automation friendly full skirt design can be sealed with Axymat or sealing film.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured with 99.9% pure virgin polypropylene
  • 384-well double notch PCR plate
  • Robotic friendly full skirt design
  • Application/Compatibility: Standard thermal cycling
  • Specifications

    Barcode No Barcode
    Certifications/Compliance RNase, DNase-free
    Format Standard
    Lid Without Lid
    Array 16 x 24
    Color Clear
    Material Polypropylene
    No. of Wells 384
    Volume (Metric) Working 40 μL
    Sterility Non-sterile
    Well Shape Round
    For Use With (Application) PCR
    Includes Double notch
    No. per Case 50/Cs.
    No. per Pack 10/Pk.
    Show More Show Less