missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 24- and 32-Well PCR Microplates, Clear, Nonsterile

Catalog No. 50400559
Change view
Click to view available options
No. of Wells:
24
32
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
50400559 24
14222303 32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50400559 Supplier Axygen™ Supplier No. PCR24C

Designed to fit a wide variety of thermal cyclers

  • Volume to brim of well: 350 μL, working volume: 300 μL, volume with AxyMat: 240 μL

Specifications

Certifications/Compliance RNase, DNase-free
Format Standard
Lid Without Lid
Array 4 x 6
Color Clear
Product Type PCR Microplate, without Lid
Material Polypropylene
No. of Wells 24
Sterility Non-sterile
Well Shape Round
For Use With (Application) PCR
No. per Case 50/Cs.
No. per Pack 10/Pk.
Show More Show Less