missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200μL Universal Pipette Tips: 200μL, Beveled
GREENER_CHOICE

Catalog No. p-4102794
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Format:
Bulk
Hinged Rack
Rack
Rack, 5 x 96
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Color Format
14222712 Non-sterile Clear Bulk
14222719 Non-sterile Clear Rack, 5 x 96
14222717 Non-sterile Clear Hinged Rack
14222718 Sterile Clear Hinged Rack
14222725 Sterile Yellow Rack
14222723 Sterile Yellow Rack
14222721 Non-sterile Yellow Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14222712 Supplier Axygen™ Supplier No. T200C
Only null left

Designed to fit a wide variety of single and multi-channel pipettes.

  • Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
  • Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Graduation marks allow a visual confirmation of pipettor draw volume
  • Presterilized tips are e-beam irradiated to prevent product degradation
  • Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
  • Also available with Maxymum recovery, which allows complete retrieval of precious samples, saves costly reagents

Order Info

Available in bulk, racked or stack rack formats.

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Bulk
Tip Style Universal, Beveled
For Use With (Equipment) Universal Fit
Material Polypropylene