missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200μL Universal Pipette Tips: 200μL, Beveled

Catalog No. 14222718
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Format:
Bulk
Hinged Rack
Rack
Rack, 5 x 96
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Color Format
14222718 Sterile Clear Hinged Rack
14222712 Non-sterile Clear Bulk
14222719 Non-sterile Clear Rack, 5 x 96
14222717 Non-sterile Clear Hinged Rack
14222725 Sterile Yellow Rack
14222723 Sterile Yellow Rack
14222721 Non-sterile Yellow Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 missing translation for 'options'
Catalog No. 14222718 missing translation for 'mfr' Axygen™ missing translation for 'supplierNo' T200CRS
Only null left

Designed to fit a wide variety of single and multi-channel pipettes.

  • Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
  • Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Graduation marks allow a visual confirmation of pipettor draw volume
  • Presterilized tips are e-beam irradiated to prevent product degradation
  • Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
  • Also available with Maxymum recovery, which allows complete retrieval of precious samples, saves costly reagents

Order Info

Available in bulk, racked or stack rack formats.

Specifications

Volume (Metric) 1 to 200 μL
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Hinged Rack
Tip Style Beveled, Universal
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene