missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 200μL Universal Pipette Tips, Beveled, Reference, Maxymum Recovery
Designed to fit a wide variety of single and multi-channel pipettes
Supplier: Axygen™ TR222CL
Description
- Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
- Maxymum recovery allows complete retrieval of precious samples, saves costly reagents
- Wide bore tips of orifice size: 0.062 in. (1.52mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
- Graduation marks allow a visual confirmation of pipettor draw volume
- Presterilized tips are e-beam irradiated to prevent product degradation
- Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
Specifications
| 1 to 200 μL | |
| Clear | |
| DNase, RNase, DNA, and PCR inhibitor Free | |
| Beveled, Low Retention, Universal | |
| Yes | |
| 10, 50, 100μL |
| Non-sterile | |
| Autoclavable | |
| Bulk | |
| Universal Fit | |
| Polypropylene |