missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 20 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
GREENER_CHOICE

Catalog No. 14222902
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Filtered
14222902 Sterile Filtered
14222875 Non-sterile Non-filtered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222902 Supplier Axygen™ Supplier No. VTF20RS
Get Up to $2,500 in Products with Purchase
View all promotions

For use with Agilent/Velocity11 automation systems

Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

 

  • Clear polypropylene
  • RNase-/DNase-free
  • Nonpyrogenic
  • Available in nonsterile or sterile

 

Specifications

Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Color Clear
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Features w/out Bottom Label
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Material Polyethylene
Show More Show Less