missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 165 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 96 Tip Format
GREENER_CHOICE

Catalog No. 14222899
Change view
Click to view available options
Includes:
w/Bottom Label
w/out Bottom Label
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
14222899 w/Bottom Label
14222900 w/out Bottom Label
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14222899 Supplier Axygen™ Supplier No. VTF165RLABS

For use with Agilent/Velocity11 automation systems

  • Tip format: 96 tip
  • Tip volume range: 20 μL to 250 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: 96LT, 8LT
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene

Specifications

Volume (Metric) 165 μL
Filtered Filtered
Sterility Sterile
Color Clear
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
Packaging SLAS Rack
Quantity 50 Racks/Cs.
Includes w/Bottom Label
For Use With (Equipment) Agilent/Velocity VPrep 8LT, Agilent/Velocity VPrep 96LT, Agilent Bravo 96LT
Material Polyethylene
Show More Show Less