missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 1000 μL Universal Pipetter Tips: Wide Bore
GREENER_CHOICE

Catalog No. p-4903293
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
14222699 Non-sterile Bulk
14222702 Non-sterile Rack
14222703 Sterile Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14222699 Supplier Axygen™ Supplier No. T1005WBC
Only null left

Compatible with most popular brands of pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

(This is the suggested replacement for T-1005-WB-C-L)

  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore

Specifications

Volume (Metric) 100 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Bulk
Tip Style Genomic/Wide Orifice, Universal
Packaging 1,000 Tips/Pk., 5 Packs/Cs.
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less