Learn More
Axygen™ 1000 μL Universal Pipetter Tips: Bevelled
Compatible with most popular brands of pipettes
Supplier:  Axygen™ T1000CRS
 View more versions of this product
Catalog No. 14222698
Only null left
Description
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
Specifications
Chemical Identifiers
100 to 1000 μL
DNase, RNase, and Pyrogen free
Universal Fit
Autoclavable
Beveled, Universal
Polypropylene
Specifications
100 to 1000 μL
Clear
DNase, RNase, and Pyrogen free
Beveled, Universal
Universal Fit
Sterile
Autoclavable
Rack
Tripled bagged
Polypropylene
Product Suggestions
Product Suggestions