Learn More
Axygen™ 1000 μL Universal Pipetter Tips: Bevelled
Compatible with most popular brands of pipettes
Supplier: Axygen™ T1000BRS
Description
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Beveled
Chemical Identifiers
| 100 to 1000 μL | |
| DNase, RNase, and Pyrogen free | |
| Universal Fit |
| Autoclavable | |
| Beveled, Universal | |
| Polypropylene |
Specifications
| 100 to 1000 μL | |
| Sterile | |
| Autoclavable | |
| Rack | |
| Universal Fit |
| Non-filtered | |
| Blue | |
| DNase, RNase, and Pyrogen free | |
| Beveled, Universal | |
| Polypropylene |