missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 20 μL Ultra Micro Tips

Catalog No. 14222756
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Format
14222756 Non-sterile Bulk
50400951 Sterile Rack
14222760 Non-sterile Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14222756 Supplier Axygen™ Supplier No. T400
Get Up to $2,500 in Products with Purchase
View all promotions

Maxymum Recovery™ allows recovery of costly samples with improved precision and accuracy

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • RNase-/DNase-free
  • Nonpyrogenic
  • Maxymum Recovery technology for ultra-low retention

Order Info

  • Bulk: 1000 tips/pack, 20 packs/case
  • Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

Specifications

Volume (Metric) 0.5 to 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model T-400
Certifications/Compliance Endotoxin freeEN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, Quality Control Tested
Clean Claims DNase, RNase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Bulk
Tip Style Micro Point
Packaging 1,000 Tips/Pk., 20 Packs/Cs.
Quantity 1000 Tips/Pk., 20 Pk./Cs.
Product Type Pipette Tips
For Use With (Equipment) Gilson Pipetman P10, Gilson Pipetman P2, Eppendorf
Product Line Maxymum Recovery
Material Polypropylene
Show More Show Less