missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 10μL Microvolume Tips

Catalog No. 14223378
Change view
Click to view available options
Volume (Metric):
0.5 to 10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14223378 0.5 to 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14223378 Supplier Axygen™ Supplier No. T300LSTKS

Axygen Universal Tips are designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, bevelled, nonsterile, sterile, non-filtered, and other features for specialized applications.

  • Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention and high yield

Specifications

Volume (Metric) 0.5 to 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Model T-300-L-STK-S
Certifications/Compliance USP 3127
Clean Claims DNase, Rnase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Format Rack
Tip Style Low Retention, Microvolume
For Use With (Equipment) Gilson Pipetman P10, Gilson Pipetman P2
Material Polypropylene
Show More Show Less