missing translation for 'onlineSavingsMsg'
Learn More

Axiom™ Wheat HD Genotyping Arrays

Catalog No. 550492
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
550492 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 550492 Supplier Thermo Fisher Scientific Supplier No. 550492

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Array Genotyping
Number of Arrays 2 x 96 arrays
Format 96-array Plate
Species Wheat
For Use With (Application) Microarray Analysis
Quantity 1 plate

For Research Use Only. Not for use in diagnostic procedures.