missing translation for 'onlineSavingsMsg'
Learn More

Axiom™ Wheat Breeder's Genotyping Array

Catalog No. 550524
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
550524 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 550524 Supplier Thermo Fisher Scientific Supplier No. 550524

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Array Genotyping
Number of Arrays 384 arrays
Format 384-array Plate
Species Wheat
Content And Storage Store in refrigerator (2°C to 8°C)
For Use With (Application) Microarray Analysis
Quantity 1 plate
Shipping Condition Room temperature

For Research Use Only. Not for use in diagnostic procedures.