Learn More
Applied Biosystems™ Axiom™ Porcine Genotyping Array
Description
Axiom Porcine Genotyping Array (Axiom_PigHDv1) for pig genotyping was designed through the Affymetrix- Expert Design Program by Dr. Martien Groenen at Wageningen University, Netherlands, and Dr. Alan Archibald at The Roslin Institute, United Kingdom.
Axiom Porcine Genotyping Array is available in 96-array format and includes 658,692 markers discovered using whole-genome sequence data from 210 animals covering a broad range of commercial and non-commercial pig breeds that were selected for their relevance to world-wide breeding programs.1 Powered by Axiom Genotyping Solution, the array offers the highest data completeness, call rates, and reproducibility of any technology available today.
This high-density array offers the power and resolution for a wide range of applications in pig breeding and genomics that include studying marker-trait association, evaluating pure lines, identifying multi-line reference populations, as well as research applications for genome-wide analysis and selective sweep analysis studies. A key benefit of using Axiom Porcine Genotyping Array is the ability to genotype samples without experiencing marker dropout or missing data, which have been observed when using other porcine genotyping products.
Features of Axiom Porcine Genotyping Array
- Expert design: developed by key opinion leaders in the porcine community
- High resolution
- 658,692 markers on the array
- Equal spacing on chromosomes
- Bias towards common variants
- 56,000 of the most informative markers from an existing in-market 60,000 marker array, allowing compatibility with previous studies
- High diversity: includes markers from a diverse set of commercially relevant breeds, including rare/traditional breeds
Applications of Axiom Porcine Genotyping Array
- Construction of high-resolution genetic maps
- Genetic improvement of pure lines
- Fine mapping of quantitative trait loci
- Developing training populations for calculating breeding values with genomic selection models
Required Products
- GeneChip Command Console Software
- GeneTitan Multi-Channel Instrument
Specifications
Specifications
| Array | Genotyping |
| Number of Arrays | 96 arrays |
| Format | 96-array Plate |
| Species | Porcine |
| For Use With (Application) | Microarray Analysis |
| Quantity | 1 plate |
Frequently Asked Questions (FAQs)
All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).
For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.
For Research Use Only. Not for use in diagnostic procedures.