Learn More
Applied Biosystems™ Axiom™ PangenomePro™ SwiftArray™ Kit
Description
The Axiom PangenomePro Array boasts the largest ethnic diversity coverage within the Axiom portfolio and enables target SNP identification, copy number detection and variant analysis, and HLA typing in a cost-effective assay with plug and play data analysis. It delivers ethnically diverse, genome-wide coverage in a high-throughput 96-array plate format, designed to support whole-genome imputation across global populations and help researchers build more representative genomic datasets. The array can serve as a core research tool for human genomics workflows including GWAS, population health initiatives, polygenic risk scores, pharmacogenomics, and drug discovery.
Features of the Axiom PangenomePro Array include:
- Marker selection that emphasizes broad imputation power across European, African, admixed American, East Asian, and South Asian populations
- Whole-genome analysis from as little as 100 ng genomic DNA with support for target SNP identification, CNV analysis, HLA and blood typing, and 'plug and play' data analysis workflows
- GWAS imputation module (~850,000 markers) across ancestral populations
- Evidence-based clinical/research markers that include references to public resources such as ACMG 73, ClinVar, NHGRI-GWAS catalog
- Pharmacogenomics coverage aligned to guideline and knowledgebase citations (e.g., CPIC, PharmGKB, PharmaADME)
- CNV analysis for fixed regions and de novo discovery across the genome
Specifications
Specifications
| Type | Human Microarray Genotyping |
| Array | Genotyping, DNA, Copy Number |
| Number of Arrays | 2 x 96 arrays |
| Format | 96-array Plate |
| Species | Human |
| No. of Samples | 2x96F |
| Quantity | 192 Reactions |
For Research Use Only. Not for use in diagnostic procedures.