missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axiom™ Ovine Genotyping Array
Specifications
Specifications
| Array | DNA |
| Number of Arrays | 384 arrays |
| Format | Array Plate |
| Content And Storage | One plate containing 384 arrays |
| For Use With (Application) | Microarray Analysis |
| Product Line | Axiom |
| Quantity | 1 plate |
For Research Use Only. Not for use in diagnostic procedures.