missing translation for 'onlineSavingsMsg'
Learn More

Axiom™ Multispecies Plant Genotyping Array, mini 96 format

Catalog No. 551231
Change view
Click to view available options
Quantity:
96 samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
551231 96 samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 551231 Supplier Thermo Fisher Scientific Supplier No. 551231

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Type Multispecies Plant Genotyping Array
Array Genotyping
Number of Arrays 96 arrays
Format Mini 96-array Plate
Species Multispecies: Cashew, Coffee, Araucaria, Cassava, Eucalyptus
Content And Storage Store in refrigerator (2°C to 8°C)
Quantity 96 samples
Shipping Condition Room temperature