missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axiom™ Genome-Wide BOS 1 Bovine Array
Specifications
Specifications
| Array | Genotyping |
| Number of Arrays | 96 arrays |
| Format | 96-array Plate |
| Species | Bovine |
| For Use With (Application) | Microarray Analysis |
| Quantity | 1 plate |
For Research Use Only. Not for use in diagnostic procedures.