missing translation for 'onlineSavingsMsg'
Learn More

Axiom™ Eucalyptus Genotyping Array (Axiom Euc72K), mini 96 format

Catalog No. 551135
Change view
Click to view available options
Quantity:
1 array
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
551135 1 array
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 551135 Supplier Thermo Fisher Scientific Supplier No. 551135

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Array Genotyping
Number of Arrays 96 arrays
Format Mini 96-array Plate
Species Eucalyptus
Content And Storage Store in refrigerator (2°C to 8°C)
Quantity 1 array
Shipping Condition Room temperature

For Research Use Only. Not for use in diagnostic procedures.