missing translation for 'onlineSavingsMsg'
Learn More

Axiom™ Canine HD Array

Catalog No. 550869
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
550869 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 550869 Supplier Thermo Fisher Scientific Supplier No. 550869

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Array DNA
Number of Arrays 96 arrays
Format 96-array Plate
Species Canine
Content And Storage Store in refrigerator (2°C to 8°C)
Product Line Axiom
Quantity 1 plate
Shipping Condition Room Temperature