missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ AX200 ESD Uncoated Gloves

Numéro de catalogue. 18139761
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Size
18139761 X-Large
18139752 Small
18139751 Medium
18139750 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 18139761 Fournisseur SHOWA™ Code fournisseur AX200XL09
Il en reste null

Electrostatic dissipative 18-gauge nylon, uncoated seamless knit glove

The new SHOWA AX200 has an 18-gauge seamless knit, with nylon static dissipative yarn. It is free of natural rubber latex and is touchscreen compatible, silicone-free and fingerprint-free. Certified under the standard EN 16350, the AX200 by SHOWA meets the requirements for electrostatic properties (ESD). This certification ensures the glove’s safe usage in ATEX zones (working area with risk of explosion and/or high flammability).

Spécifications

Accelerator Free Yes
Applications Industrial
Coating Material Uncoated
Color Light Gray
Disposable Reusable
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Nylon
Powdered Powder Free
Product Type ESD Gloves
Size X-Large
Sterility Non-sterile
Thickness (English) 18 gauge
Thickness (Metric) 0.8 mm
Afficher plus de résultats Afficher moins de résultats