missing translation for 'onlineSavingsMsg'
Learn More

Avobenzone, 97%, Spectrum™ Chemical

Catalog No. p-7156970
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600816 5 g Amber Glass Bottle
18600817 25 g Amber Glass Bottle
18600818 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18600816 Supplier Spectrum Chemical Mfg Cor Supplier No. A26825GM
Only null left

A2682, 70356-09-1, C20H22O3

Spectrum™ Chemical Avobenzone is an oil soluble ingredient used in sunscreen products to absorb the full spectrum of UVA rays. Its ability to absorb ultraviolet light over a wider range of wavelengths than many organic sunscreen agents has led to its use in many commercial preparations marketed as ′broad spectrum′ sunscreens. Avobenzone has an absorption maximum of 357 nm. It appears as an odorless, yellowish solid.

Chemical Identifiers

CAS 70356-09-1
Molecular Formula C20H22O3
Molecular Weight (g/mol) 310.39
MDL Number MFCD00210252
InChI Key XNEFYCZVKIDDMS-UHFFFAOYSA-N
IUPAC Name 1-(4-tert-butylphenyl)-3-(4-methoxyphenyl)propane-1,3-dione
SMILES COC1=CC=C(C=C1)C(=O)CC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 97%
Grade Reagent