missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avobenzone, 97%, Spectrum™ Chemical
Description
Spectrum™ Chemical Avobenzone is an oil soluble ingredient used in sunscreen products to absorb the full spectrum of UVA rays. Its ability to absorb ultraviolet light over a wider range of wavelengths than many organic sunscreen agents has led to its use in many commercial preparations marketed as ′broad spectrum′ sunscreens. Avobenzone has an absorption maximum of 357 nm. It appears as an odorless, yellowish solid.
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Quantity | 100 g |
| Percent Purity | 97% |
| Grade | Reagent |
| Physical Form | Crystalline Powder or Lumps |
| Chemical Name or Material | Avobenzone |