Learn More
Research Products Corporation Autoradiography Cassette, 5 x 7 Inches
Catalog No. 50212720
Encompass
RPI film cassettes include innovative features designed for autoradiography procedures and provide superior performance in laboratory applications. Cassettes withstand repeated exposure to temperatures down to -80°C. Durable hinges and latches operate freely without freezing up. Convenient push button latches are easy to locate and use in a darkroom. High strength aluminum construction with a non-chip finish for comfortable handling even at low temperatures. Integral light lock prevents stray light from entering cassette and damaging film. High resiliency foam pressure pad ensures maximum contact between gel and film. All cassettes undergo intensive lightproof testing as part of a stringent quality control procedure.
Catalog No. 50212720 Supplier Research Products Corporation Supplier No. RPI 420057
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RPI film cassettes include innovative features designed for autoradiography procedures and provide superior performance in laboratory applications. Cassettes withstand repeated exposure to temperatures down to -80°C. Durable hinges and latches operate freely without freezing up. Convenient push button latches are easy to locate and use in a darkroom. High strength aluminum construction with a non-chip finish for comfortable handling even at low temperatures. Integral light lock prevents stray light from entering cassette and damaging film. High resiliency foam pressure pad ensures maximum contact between gel and film. All cassettes undergo intensive lightproof testing as part of a stringent quality control procedure.