Learn More
Labscientific Inc. AUTORAD BLUE FILM 8X10 100/PK
Catalog No. NC9648989
Encompass
Western Blotting, Sequencing, Chemiluminescent Autoradiography (ecl), and Gel Shift Analysis.Superior Clarity. Very Low Background Fogging. Publishable Autoradiographs. Compatible with all manual and automatic development. Guaranteed 30 month Expiration Date. 8″ x 10″ (20.4 x 25.5 cm) Pack of 100
Catalog No. NC9648989 Supplier Labscientific Inc. Supplier No. XAR ALF 2025
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Western Blotting, Sequencing, Chemiluminescent Autoradiography (ecl), and Gel Shift Analysis.Superior Clarity. Very Low Background Fogging. Publishable Autoradiographs. Compatible with all manual and automatic development. Guaranteed 30 month Expiration Date. 8″ x 10″ (20.4 x 25.5 cm) Pack of 100