Learn More
Labscientific Inc. Autoradiographic film product. Dimensions: 5 x7 in. (13x18cm), sharp contrast, high resolution, blotting and sequence autoradiography, 100/pack
Catalog No. NC9648990
Encompass
Western Blotting, Sequencing, Chemiluminescent Autoradiography (ecl), and Gel Shift Analysis. Superior Clarity. Very Low Background Fogging. Publishable Autoradiographs. Compatible with all manual and automatic development. The sharp contrast and high resolution is ideal for sequencing especially in the Lifescience Fields such as: Western Blotting, DNA and Microbiology testing. The film features low patient dose and excellent image quality for blotting & sequence autoradiography. Optimized for chemiluminescence. Specialized film for: Research labs, forensic labs, laboratory testing, and research hospitals. Guaranteed 30 month Expiration Date. Dimensions: 5 x 7 (13 x 18 cm). 100/pack.
Catalog No. NC9648990 Supplier Labscientific Inc. Supplier No. XAR ALF 1318
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Western Blotting, Sequencing, Chemiluminescent Autoradiography (ecl), and Gel Shift Analysis. Superior Clarity. Very Low Background Fogging. Publishable Autoradiographs. Compatible with all manual and automatic development. The sharp contrast and high resolution is ideal for sequencing especially in the Lifescience Fields such as: Western Blotting, DNA and Microbiology testing. The film features low patient dose and excellent image quality for blotting & sequence autoradiography. Optimized for chemiluminescence. Specialized film for: Research labs, forensic labs, laboratory testing, and research hospitals. Guaranteed 30 month Expiration Date. Dimensions: 5 x 7 (13 x 18 cm). 100/pack.