missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec™ Autopsy Saw Blades, SuperCut, Compatible with Stryker Saw
Mopec's exclusive titanium coating assures longer and sharper blade life. Fits all saws with radial mount arbors.
Supplier: Mopec™ BD114
Description
- Dimensions: 2.5 in. Diameter (64mm)
- Material: 420 Stainless steel with Titanium Nitride coating for added strength
- Fits Mopec Autopsy Saw (excludes Mopec Autopsy Saw 5000) and Stryker™ 810 Saw
Chemical Identifiers
| Titanium Nitride Coating for added strength | |
| Stainless Steel (420) | |
| 2.5 in. |
| Autopsy Saw Blades, SuperCut, Compatible with Stryker Saw | |
| Fits Mopec Autopsy Saw (excludes Mopec Autopsy Saw 5000) and Stryker™ Saw |
Specifications
| Yes | |
| Autopsy Saw Blades, SuperCut, Compatible with Stryker Saw | |
| Fits Mopec Autopsy Saw (excludes Mopec Autopsy Saw 5000) and Stryker™ Saw | |
| 2.5 in. |
| Titanium Nitride Coating for added strength | |
| Stainless Steel (420) | |
| Round Blade |