Learn More
Mopec™ Autopsy Saw Blades, SuperCut, Compatible with Stryker Saw
Catalog No. BD113
Change view
Click to view available options
Arbor Included:
No
Yes
Style:
Round Blade
Section Blade
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arbor Included Style
BD113 Yes Section Blade
BD114 Yes Round Blade
BD115 No Section Blade
BD116 No Round Blade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BD113 Supplier Mopec™ Supplier No. BD113
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mopec's exclusive titanium coating assures longer and sharper blade life. Fits all saws with radial mount arbors.
  • Dimensions: 2.5 in. Diameter (64mm)
  • Material: 420 Stainless steel with Titanium Nitride coating for added strength
  • Fits Mopec Autopsy Saw (excludes Mopec Autopsy Saw 5000) and Stryker™ 810 Saw
Specifications
Arbor Included Yes
Coating Titanium Nitride Coating for added strength
Description Autopsy Saw Blades, SuperCut, Compatible with Stryker Saw
Material Stainless Steel (420)
For Use With (Equipment) Fits Mopec Autopsy Saw (excludes Mopec Autopsy Saw 5000) and Stryker™ Saw
Style Section Blade
Diameter (English) Blade 2.5 in.