missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Autopsy Dissecting Knives, 15 mm Wide

All-purpose autopsy dissecting knife with a grooved handle to provide a secure grip.

Supplier:  Mopec™ AH011

 View more versions of this product

Catalog No. 22445385


Only null left

Description

Description

  • Made of standard grade stainless steel
  • Cutting edge is ridged, tapered blade with pointed tip
Specifications

Chemical Identifiers

Autopsy Dissecting Knife
15 mm
Reusable
No

Specifications

Autopsy Dissecting Knife
15 mm
No
200 mm
Reusable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions