missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
GREENER_CHOICE

Catalog No. 21403428
Change view
Click to view available options
Volume (Metric):
50 μL
80 μL
100 μL
150 μL
200 μL
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Filtered Fit for Purpose
21403428 50 μL Sterile Filtered Sterile and PCR Ready
21403426 50 μL Sterile Non-filtered Sterile
21403427 50 μL Non-sterile Non-filtered Certified Quality
21403431 80 μL Sterile Filtered Sterile and PCR Ready
21403429 100 μL Sterile Non-filtered Sterile
02682273 100 μL Non-sterile Non-filtered Certified Quality
21403434 150 μL Sterile Filtered Sterile and PCR Ready
21403432 200 μL Sterile Non-filtered Sterile
21403433 200 μL Non-sterile Non-filtered Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 21403428 Supplier Thermo Scientific™ Supplier No. 906021

Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.

  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 50 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius
Product Line Tecan
Fit for Purpose Sterile and PCR Ready