Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Supplier:  Thermo Scientific™ 901251
 View more versions of this product
Catalog No. 21-402-318
Only null left
Description
Description
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
Specifications
Specifications
20 μL
Sterile
Not Autoclavable
Free of RNase, DNase, DNA, ATP, and endotoxin
Conductive
960 Tips
Tecan
Non-filtered
Clear
ISO 9001 and ISO 13485
Rack
Reload Insert, 96 tips/insert, 10 inserts/pack
Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Sterile
Product Suggestions
Product Suggestions