missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
GREENER_CHOICE

Catalog No. 21402345
Change view
Click to view available options
Volume (Metric):
175 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21402345 175 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21402345 Supplier Thermo Scientific™ Supplier No. 9030111

Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms increases pipetting perfomrance through the superior tip straightness and low %CV of these pipette tips.

Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.

  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 175 μL
Filtered Filtered
Sterility Sterile
Color Black
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims RNase, DNase, DNA, ATP and endotoxin free
Format Rack
Tip Style Specialty
Quantity 96 Tips/Insert, 10 Inserts/Pk.
For Use With (Equipment) Cavro, MiniPrep, Tecan Freedom EVO, Tecan Genesis
Show More Show Less