missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
GREENER_CHOICE

Catalog No. 21402329
Change view
Click to view available options
Volume (Metric):
20 μL
130 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility Fit for Purpose
21402329 200 μL Non-filtered Non-sterile Certified Quality
21402324 20 μL Filtered Sterile Sterile and PCR Ready
21402325 20 μL Non-filtered Sterile Sterile
21402326 20 μL Non-filtered Non-sterile Certified Quality
21402327 130 μL Filtered Sterile Sterile and PCR Ready
21402328 200 μL Non-filtered Sterile Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 21402329 Supplier Thermo Scientific™ Supplier No. 917262

Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Extended Length
For Use With (Equipment) Beckman Coulter Biomek 1000/2000
Product Line Beckman
Fit for Purpose Certified Quality