missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
GREENER_CHOICE

Catalog No. 21402192
Change view
Click to view available options
Volume (Metric):
8 μL
10 μL
20 μL
40 μL
50 μL
130 μL
175 μL
250 μL
Filtered:
Filtered
Non-filtered
Tip Style:
Extended Length
Low Retention
Standard
Wide Bore
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Tip Style Fit for Purpose
21402192 20 μL Non-filtered Standard Certified Quality
14754342 8 μL Filtered Standard Sterile and PCR Ready
14754340 10 μL Non-filtered Standard Sterile
14754341 10 μL Non-filtered Standard Certified Quality
21402650 40 μL Filtered Standard Sterile and PCR Ready
21402651 50 μL Non-filtered Standard Sterile
21402652 50 μL Non-filtered Standard Certified Quality
21402190 20 μL Filtered Standard Sterile and PCR Ready
21402638 20 μL Filtered Low Retention Sterile and PCR Ready
21402191 20 μL Non-filtered Standard Sterile
21402636 20 μL Non-filtered Low Retention Sterile
21402637 20 μL Non-filtered Low Retention Certified Quality
21402193 130 μL Filtered Standard Sterile and PCR Ready
21402641 130 μL Filtered Low Retention Sterile and PCR Ready
14754324 130 μL Filtered Wide Bore Sterile and PCR Ready
21402194 250 μL Non-filtered Standard Sterile
21402639 250 μL Non-filtered Low Retention Sterile
21402360 250 μL Non-filtered Wide Bore Sterile
21402195 250 μL Non-filtered Standard Certified Quality
21402640 250 μL Non-filtered Low Retention Certified Quality
14754325 250 μL Non-filtered Wide Bore Certified Quality
21402237 50 μL Filtered Extended Length Sterile
21402224 175 μL Filtered Standard Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Catalog No. 21402192 Supplier Thermo Scientific™ Supplier No. 918262

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Standard
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 4800 tips
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Certified Quality
Show More Show Less