missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head

Catalog No. p-6858103
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Filtered Fit for Purpose
14754375 Sterile Filtered Sterile and PCR Ready
21935261 Sterile Non-filtered Sterile
21935262 Non-sterile Non-filtered Certified Quality
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 14754375 Supplier Thermo Scientific™ Supplier No. 935021
Only null left

Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)

Specifications

Volume (Metric) 30 μL
Filtered Filtered
Sterility Sterile
Color Clear
Tip Style Low Retention, Extended Length
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile and PCR Ready