missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Automatic Burettes

Catalog No. 22261822
Change view
Click to view available options
Capacity (Metric):
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22261822 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22261822 Supplier Bel-Art™ Supplier No. H379180109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Self zeroing glass burettes.

  • With easy-to-read blue graduations against white background
  • Delivery end fitted into plastic push button stopcock which permits drop by drop delivery
  • Small screw clamp adjusts drop speed as needed for titration
  • Polyethylene filling tube extends from top of burette into bottle reservoir
  • Polyethylene plastic bottle, screw cap and round base
  • Class B graduation accuracy

Specifications

Accuracy Class B
Autoclavable Not Autoclavable
Closure Type Screw Cap
Color Natural
Diameter (Metric) Outer 7.62 cm
Capacity (Metric) 10 mL
Length (Metric) 21 cm
For Use With (Equipment) Stirplate
Material Glass
For Use With (Application) Titration, Reagent Dispensing
Includes Bottle, Round Base, Screw Cap
Scale Blue
Stopcock No
Tolerance Class B
Warranty None
Volume (Metric) Reservoir 500 mL
Type Automatic Buret
Class Class B
Show More Show Less