missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Automatic 8-Cell Changer

Catalog No. BC80710008
Change view
Click to view available options
Type:
Ambient Multi-cell Holder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
BC80710008 Ambient Multi-cell Holder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BC80710008 Supplier Biochrom Supplier No. 80710008

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Automatic linear Cell Changer with 8 position ambient multicell holder. For use with Ultrospec 7000/8000/9000 Spectrophotometer.

  • Automatic linear cell changer with 8 position ambient multicell holder
  • Accepts 10 mm pathlength cuvettes
  • Includes 10 mm single cell holder for reference beam
  • Easy to install
  • For use with Ultrospec 7000/8000/9000 Spectrophotometer

Specifications

Compatibility Ultrospec 7000/8000/9000 Spectrophotometer
Type Ambient Multi-cell Holder
Disposable Reusable
Pathlength 10 mm
Shape Rectangular