missing translation for 'onlineSavingsMsg'
Learn More

DWK LIFE SCIENCES™ Autoclaving Box for 10 mL Pipette Tips

Catalog No. 02912682
Change view
Click to view available options
Holds:
Pipette Tips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
02912682 Pipette Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02912682 Supplier DWK LIFE SCIENCES™ Supplier No. W835150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Holds 24 pipette tips (10 mL) for autoclave cleaning and sterilizing.

  • Pipette tip cleaning equipment

Specifications

Autoclavable Autoclavable
Color Natural
Holds Pipette Tips
Material Polypropylene
Positions 24
Shape Cubical
Stackable No
Sterility Non-sterile
For Use With (Equipment) 10 mL Pipette Tips
Product Type Empty Tip Box
Array 6 x 4
Show More Show Less