missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Waste Bags: Printed

Printed style.

Supplier:  Fisherbrand™ 1826E

 View more versions of this product

Catalog No. 01826E


Only null left

Description

Description

Printed style bags

  • Maximum temperature, 140°C (285°F); for use at 134°C (274°F)
  • 2mil thick
  • Supplied with autoclavable elastic steam-venting closures

Compliance

Graphics on printed styles conform to OSHA™ standard 29CFR1910.1030.

Specifications

Chemical Identifiers

Autoclavable
Elastic
Polypropylene
2 mil
OSHA, 29 CFR1910.1030
Printed in English and Spanish
140°C

Specifications

Autoclavable
Elastic
Printed in English and Spanish
140°C
48 x 37 in. (122 x 94 cm)
37 in.
122 cm
OSHA, 29 CFR1910.1030
Clear
Polypropylene
2 mil
48 in.
44 gal.
94 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions