missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Autoclavable Biohazardous Waste Bags

Autoclavable Biohazardous Waste Bags are built to withstand the rigors of Steam Sterilization while meeting the requirements for collection, storage, transport and processing of biohazardous waste within a facility.

Supplier:  Fisherbrand™ 1831C

Catalog No. 01831C

Only null left
Explore available promotions

Description

Description

  • Single-use Biohazard waste bag
  • ASTMD1709 Impact Testing
Specifications

Specifications

Autoclavable
15 L
Twist Tie
Single-use
Non-sterile
4 mil
48 in.
38 in.
Autoclavable Bags
44 gal.
ASTMD1709 Impact Testing
Red
PET, Polypropylene Film
0.1 mm
38 x 48 in.
122 cm
96.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions