missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Autocamtide-2-related inhibitory peptide, myristoylated
Description
Autocamtide-2-related inhibitory peptide, myristoylated is a CaM kinase II inhibitor; enhanced cell permeable derivative of Autocamtide-2-related inhibitory peptide (Cat No. 1688). Pretreatment blocks reinstatement of morphine-seeking behavior in vivo.
Specifications
Specifications
| Purity | 0.95 |
| CAS | 201422-04-0 |
| Content And Storage | Store at -20°C |
| Description | CaM Kinase Inhibitors |
| Molecular Weight (g/mol) | Observed M.W.: 1708.12 |
| Quantity | 1 mg |
| Structural Form | C78H142N22O20 |