missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Auramine O, pure, biological stain

Catalog No. AC227520500
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC227520500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC227520500 Supplier Thermo Scientific Chemicals Supplier No. 227520500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2465-27-2 | C17H22ClN3 | 303.834 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2465-27-2
Molecular Formula C17H22ClN3
Molecular Weight (g/mol) 303.834
MDL Number MFCD00012484
InChI Key KSCQDDRPFHTIRL-UHFFFAOYSA-N
Synonym Basic Yellow 2, 4, 4'-(Imidocarbonyl)bis(N, N-dimethylaniline) monohydrochloride, C.I. 41000
PubChem CID 17170
ChEBI CHEBI:51876
IUPAC Name 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl

Specifications

Melting Point >250.0°C
Dye Content 60% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Color Ochre/Yellow to Yellow
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in dimethylformamide, acetone and ethanol
Quantity 50 g
Physical Form Powder
Formula Weight 303.83
Grade Pure
Chemical Name or Material Auramine O
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Suspected of causing cancer.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only