Learn More
Thermo Scientific Chemicals Auramine O, 80% dye content
Fluorescent stain for acid-resistant bacteria in sputum | CAS: 2465-27-2 | C17H22ClN3 | 303.834 g/mol
$89.32 - $160.12
Chemical Identifiers
| CAS | 2465-27-2 |
|---|---|
| Molecular Formula | C17H22ClN3 |
| Molecular Weight (g/mol) | 303.834 |
| MDL Number | MFCD00012484 |
| InChI Key | KSCQDDRPFHTIRL-UHFFFAOYSA-N |
| Synonym | C.I. 41000 |
| PubChem CID | 17170 |
| ChEBI | CHEBI:51876 |
| IUPAC Name | 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride |
| SMILES | CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6034314
|
Thermo Scientific Chemicals
J6034314 |
25 g |
Each for $89.32
|
|
|||||
|
AAJ6034318
|
Thermo Scientific Chemicals
J6034318 |
50 g |
Each for $160.12
|
|
|||||
Description
Fluorescent stain for acid-resistant bacteria in sputum
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2465-27-2 | |
| 303.834 | |
| KSCQDDRPFHTIRL-UHFFFAOYSA-N | |
| 17170 | |
| 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride |
| C17H22ClN3 | |
| MFCD00012484 | |
| C.I. 41000 | |
| CHEBI:51876 | |
| CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl |
Specifications
| 250°C (decomposition) | |
| C17H22ClN3 | |
| UN2811 | |
| C.I. 41000 | |
| KSCQDDRPFHTIRL-UHFFFAOYSA-N | |
| 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride | |
| 17170 | |
| 303.84 | |
| 25 g | |
| Auramine O, 80% dye content |
| 2465-27-2 | |
| MFCD00012484 | |
| 4030061 | |
| Soluble in water or ethanol | |
| CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl | |
| 303.834 | |
| CHEBI:51876 | |
| Yellow | |
| Powder |
Safety and Handling
GHS H Statement
H311-H351-H302
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P361-P363-P501c
H302-H311-H351
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 219-567-2
RTECSNumber : BY3675000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only