Learn More
Eisco™ Augite (Pyroxene Mineral) Specimen
Catalog No. S24674
Change view
Click to view available options
Quantity:
12/Pk.
Catalog No. Quantity
S24674 12/Pk.
1 options
Catalog No. S24674 Supplier Eisco™ Supplier No. ESNG0011PK12
Augite (Pyroxene Mineral) Specimen suitable for use in classroom identification lab exercises and for study on 2-plane, 90° cleavage.
  • For lessons on Bowen's reaction series
  • For mineral identification and physical property exercises
  • Handy visual aid during lessons on Bowen's reaction series as well
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Mineral
For Use With (Application) Study on Bowen's reaction series
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Augite