Description
- Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer
- Helps maintain cleanliness of the fluidics lines to minimize contamination or carryover
- Formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system
- It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer
- The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system
- Solution is stable for 30 days after the bottle is opened
- The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality
- Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer
Specifications
| Store at room temperature. | |
| 1 L | |
| Attune |
| Wash Solution | |
| Attune™ Acoustic Focusing Cytometer |
For Research Use Only. Not for use in diagnostic procedures.