Description
Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer to maintain cleanliness of the fluidics lines, minimizing contamination or carryover.Maintains System Cleanliness
Attune™ Wash Solution has been formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system. It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer.
Convenient
The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The wash solution is stable for 30 days after the bottle is opened.
High Quality
The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
Chemical Identifiers
| Store at room temperature. | |
| Wash Solution | |
| Attune™ Acoustic Focusing Cytometer | |
| Attune |
Specifications
| Attune | |
| Store at room temperature. | |
| 250 mL |
| Wash Solution | |
| Attune™ Acoustic Focusing Cytometer |
For Research Use Only. Not for use in diagnostic procedures.